missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PTFE Bottles

For use with aggressive or valuable reagents

$1,000.77

Spécifications

Material PTFE
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Height (Metric) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Height (Metric) Prix Quantity & Availability  
AA38115KT
Afficher les documents Détails de la promotion
Thermo Scientific™
38115KT
1 L 185 mm
chaque for $1,000.77
Il en reste null
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Inert and leak-free
  • Flexible and with PTFE cap
Spécifications

Spécifications

PTFE
Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Ambient temperatures

FDS
Documents

Documents