missing translation for 'onlineSavingsMsg'
Learn More

Psoralen, 97%, Thermo Scientific Chemicals

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

Supplier:  Thermo Scientific Chemicals J60042ME

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ60042ME

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Intercalates into DNA; photochemical reagent for the investigation of nucleic acid structure

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Psoralen
163°C to 164°C
C11H6O3
500 mg
14,7928
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
7H-furo[3,2-g]chromen-7-one
6199
186.16
Solid
66-97-7
White
MFCD00010520
152784
psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
O=C1OC2=CC3=C(C=CO3)C=C2C=C1
186.17
CHEBI:27616
97%

Identifiants chimiques

66-97-7
186.17
ZCCUUQDIBDJBTK-UHFFFAOYSA-N
6199
7H-furo[3,2-g]chromen-7-one
C11H6O3
MFCD00010520
psoralen, ficusin, 7h-furo 3,2-g chromen-7-one, furocoumarin, psoralene, 7h-furo 3,2-g 1 benzopyran-7-one, psorline-p, 6,7-furanocoumarin, furo 3,2-g chromen-7-one, furo 3,2-g coumarin
CHEBI:27616
O=C1OC2=CC3=C(C=CO3)C=C2C=C1
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 200-639-7

missing translation for 'rtecsNumber' : LV0944000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only