Learn More
Enzo Life Sciences PROTEOSTAT Aggresome detection kit (1x25tests) DFS Item
Supplier:  Enzo ENZ510350025
Robust, quantitative detection of aggresomes relevant to the study of neurodegenerative disease, liver disease and toxicology. Aggresomes are inclusion bodies of aggregated, misfolded proteins that form when the ubiquitin-proteasome protein-degradation machinery is overwhelmed. Typically, aggresomes form in response to cellular stress and provide a cytoprotective mechanism by isolating misfolded proteins, leading ultimately to their clearance by autophagy. Protein aggregate formation is furthermore a hallmark of a variety of human diseases, such as Alzheimer's, Parkinson's, Amyotrophic lateral sclerosis or alcoholic liver disease.The PROTEOSTAT.PROTEOSTAT.. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection. Contents: PROTEOSTATAggresome detection reagent. Hoechst 33342 Nuclear stain. Proteasome Inhibitor (MG-132). 10X Assay Buffer
Numéro de catalogue. NC1190227
Il en reste null
Découvrez plus d'offres spéciales