Learn More
Enzo Life Sciences Proteasome 20S beta1i subunit monoclonal antibody (LMP2 - 13) (100 l) DFS Item
Supplier:  Enzo BMLPW88400100
Clone: LMP2-13. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human 1i protein. UniProt: P28065 (human), P28077 (rat). Source: Purified from ascites. Species Reactivity: Human, Rat. Specificity: Recognizes the 1i subunit of proteasome 20S. Applications: IHC, WB. Purity: Protein G-affinity purified ascites. Formulation: Liquid. In PBS containing 10mM sodium azide.
Numéro de catalogue. NC1528217
Il en reste null
Découvrez plus d'offres spéciales