missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 50 Coveralls, Open Wrists and Ankles
Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.
Supplier: DuPont™ NB120SWHXL002500
Avertissement:
Should not be used around heat, flame, sparks or potentially flammable or explosive environments.
Description
- Made with a microporous film that is laminated to a spundbonded polypropylene substrate
- Front zipper closure
- Open wrists
- Open ankles
- Serged seams
- Collar
- White
- Thickness: 10 mils
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Spécifications
Single-use Coveralls | |
Unisex | |
White | |
Open | |
Collared | |
Front Zipper | |
Non-sterile | |
NB120S | |
28.25 in. | |
10 mil | |
35 in. | |
27 in. | |
5.22 to 6.16 ft. | |
25/Cs. |
X-Large | |
Non-woven Fabric with Microporous Film | |
Single-use | |
Open | |
Unhooded | |
Serged | |
Laydown | |
40.75 to 44.25 | |
71.75 cm | |
Janitorial, Sanitation, General Industrial, Maintenance | |
88.90 cm | |
68.58 cm | |
1.73 to 1.88 m |