missing translation for 'onlineSavingsMsg'
Learn More

Promega PureYield™ Plasmid Midiprep System Solutions

Reagent used in PureYield™ Plasmid DNA Purification Protocol Midiprep System.

$99.92 - $108.48

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) PureYield™ plasmid Midiprep system
View More Specs

Products 3
missing translation for 'productCode' missing translation for 'brand' Quantity Product Type Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Quantity Product Type Price Quantity & Availability  
PRA1485
Encompass
missing translation for 'viewDocuments' Promotion Details
Promega
A1485
500 mL Neutralization Solution
Each for $108.48
Only null left
 
PRA7115
Encompass
missing translation for 'viewDocuments' Promotion Details
Promega
A7115
315 mL Cell Resuspension Solution
Each for $99.92
Only null left
 
PRA7125
Encompass
missing translation for 'viewDocuments' Promotion Details
Promega
A7125
315 mL Cell Lysis Solution
Each for $103.72
Only null left
 
Description

Description

  • Mfr. No. A1485 - Neutralization Solution
  • Mfr. No. A7115 - Cell Resuspension Solution
  • Mfr. No. A7125 - Cell Lysis Solution
Specifications

Specifications

15°C to 30°C
PureYield™ plasmid Midiprep system
Videos
SDS
Documents

Documents