Learn More
Promega PureYield™ Plasmid Midiprep System Solutions
Reagent used in PureYield™ Plasmid DNA Purification Protocol Midiprep System.
Supplier:  Promega A7125
Encompass
 View more versions of this product
Catalog No. PRA7125
Only null left
Description
Description
  • Reagent Used in Plasmid DNA Isolation Protocols
  • Cat.# A7122 is a component of the Wizard(R) Plus DNA Purification Systems
  • Cat.# A7125 is a component of the PureYield™ Plasmid Midiprep System
  • Cat.# A7124 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
Specifications
Chemical Identifiers
15°C to 30°C
PureYield™ plasmid Midiprep system
Specifications
15°C to 30°C
PureYield™ plasmid Midiprep system
Cell Lysis Solution
315 mL
Product Suggestions
Product Suggestions