missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PureYield™ Plasmid Midiprep System Solutions
Reagent used in PureYield™ Plasmid DNA Purification Protocol Midiprep System.
Supplier: Promega A1485
Description
- Reagents Used in DNA Plasmid DNA Purification Protocols
- Cat.# A7131 is a component of the Wizard MagneSil Tfx™ and Wizard(R) Plus DNA Purification Systems
- Cat.# A7132 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
- Cat.# A1485 is a component of the PureYield™ Plasmid Midiprep System
- Cat.# A1488 is a component of the Wizard(R) SV 96 Plasmid DNA Purification System
Chemical Identifiers
| 15°C to 30°C | |
| PureYield™ plasmid Midiprep system |
Specifications
| 15°C to 30°C | |
| PureYield™ plasmid Midiprep system | |
| Neutralization Solution | |
| 500 mL |