missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega PureYield™ Plasmid Midiprep System Solutions
Reagent used in PureYield™ Plasmid DNA Purification Protocol Midiprep System.
Supplier: Promega A7115
Description
- Reagent Used in Plasmid DNA Purification Protocols
- Cat.# A7112 is a component of the Wizard(R) Plus DNA Purification Systems
- Cat.# A7115 is a component of the PureYield™ Plasmid Midiprep System
- Cat.# A7113 and A7118 are components of the Wizard(R) SV 96 Plasmid DNA Purification System
- Cat.# A7114 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
Chemical Identifiers
| 15°C to 30°C | |
| PureYield™ plasmid Midiprep system |
Specifications
| 15°C to 30°C | |
| PureYield™ plasmid Midiprep system | |
| Cell Resuspension Solution | |
| 315 mL |