missing translation for 'onlineSavingsMsg'
Learn More

Promega MagneHis™ Protein Purification System DFS Item

The MagneHis Protein Purification System provides a magnetic-based, rapid method for the purification of polyhistidine- or HQ-tagged, expressed proteins.

$409.77 - $1698.39

Specifications

Kit Contents 50mL MagneHis binding/Wash Buffer, 15mL FastBreak™ cell lysis reagent, DNase I, 1 vial lyophilized, 10mL MagneHis Elution Buffer, 2mL MagneHis Ni-particles
Content And Storage 2°C to 10°C
For Use With (Application) Rapid and reliable purification of Polyhistidine-tagged Proteins from 1mL to 1L of bacterial culture
Type Magnetic Bead Protein Purification System
View More Specs

Includes: MagneHis binding/wash buffer, 50mL; FastBreak™ cell lysis reagent, 15mL; DNase I, lyophilized, 1 vial; MagneHis elution buffer, 10mL; MagneHis Ni-particles, 2mL

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-V8500 DFS Item
View Documents
Promega
V8500
65 Reactions
Each for $668.03
Only null left
 
PR-V8550 DFS Item
View Documents
Promega
V8550
325 Reactions
Each for $1,698.39
Only null left
 
PR-V8560 DFS Item
View Documents
Promega
V8560
2 mL
Each for $409.77
Only null left
 
PR-V8565 DFS Item
View Documents
Promega
V8565
10 mL
Each for $872.61
Only null left
 
Description

Description

  • Magnetic Purification of Polyhistidine- or HQ-tagged Proteins from E. coli or Mammalian Cell Lysates
  • No centrifugation or vacuum required after cell lysis
  • Particles are compatible with a variety of common buffers
  • Binding capacity up to 1mg of polyhistidine-tagged protein per 1mL of particles

  • U.S. Pat. Nos. 7,112,552; 7,304,015; 7,304,016; 7,354,750; 6,174,704; and 7,319,021 and other patents and patents pending.
  • Certain applications of this product are covered by patents issued and applicable in certain countries. Because purchase of this product does not include a license to perform any patented application, users of this product may be required to obtain a patent license depending upon the particular application and country in which the product is used.

Specifications

Specifications

50mL MagneHis binding/Wash Buffer, 15mL FastBreak™ cell lysis reagent, DNase I, 1 vial lyophilized, 10mL MagneHis Elution Buffer, 2mL MagneHis Ni-particles
2°C to 10°C
Rapid and reliable purification of Polyhistidine-tagged Proteins from 1mL to 1L of bacterial culture
Magnetic Bead Protein Purification System
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'