missing translation for 'onlineSavingsMsg'
Learn More

Promega JM109 Competent Cells

Numéro de catalogue. PRL1001
Encompass
Change view
Click to view available options
Quantity:
5 x 0.2 mL
Concentration:
>107cfu/μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Concentration
PRL1001 5 x 0.2 mL >107cfu/μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PRL1001 Fournisseur Promega Code fournisseur L1001
Il en reste null

JM109 is a K strain that is recA- and endA- to minimize recombination and improve the quality of plasmid DNA. In addition, the cells carry the F' episome, which allows blue/white screening.

  • Ready-to-Use Competent E. coli Cells for Recombinant Cloning and Subcloning
  • Cells are ready to use—no preparation time necessary
  • Transformation efficiencies guaranteed
  • No worries about undesirable recombination events (recA- mutation), and no nuclease carryover in miniprep DNA (endA- mutation)

Spécifications

Concentration >107cfu/μg
Content And Storage Store at -70°C.
For Use With (Application) Transformation of unmethylated DNA derived from PCR, cDNA and many other sources
Includes JM109 Competent Cells (subcloning efficiency) and Control DNA
pH 7
Product Type Competent Cells
Quantity 5 x 0.2 mL