missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoScript™ Reverse Transcriptase
GoScript Reverse Transcriptase utilizes M-MLV and state-of-the-art buffer technology designed for qPCR to deliver robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors.
$230.59 - $891.57
Spécifications
| Content And Storage | −30 to −10°C |
|---|---|
| Description | GoScript Reverse Transcriptase |
| Form | Liquid |
| pH | 7 |
| For Use With (Application) | cDNA Synthesis, RT-PCR |
Comprend: GoScript Reverse Transcriptase, GoScript 5X Reaction Buffer, MgCl2
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
PRA5003
|
Promega
A5003 |
100 Reactions |
chaque for $230.59
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
PRA5004
|
Promega
A5004 |
500 Reactions |
chaque for $891.57
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Reliable cDNA Synthesis for Sensitive Detection of Expression
- Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
- Sensitive transcription of both high-copy and low-copy messages
- Transcribes short and long transcripts; processes through secondary structure
Spécifications
| −30 to −10°C | |
| Liquid | |
| cDNA Synthesis, RT-PCR |
| GoScript Reverse Transcriptase | |
| 7 | |
| GoScript reverse transcriptase, GoScript 5X reaction buffer, MgCl2 |
U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.