missing translation for 'onlineSavingsMsg'
Learn More

Promega GoScript™ Reverse Transcriptase

Catalog No. PRA5003
Encompass
Change view
Click to view available options
Quantity:
100 Reactions
500 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA5003 100 Reactions
PRA5004 500 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA5003 Supplier Promega Supplier No. A5003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

GoScript Reverse Transcriptase, GoScript 5X Reaction Buffer, MgCl2

GoScript Reverse Transcriptase utilizes M-MLV and state-of-the-art buffer technology designed for qPCR to deliver robust, reliable cDNA synthesis of a full range of rare and abundant transcripts, even in the presence of inhibitors.

  • Reliable cDNA Synthesis for Sensitive Detection of Expression
  • Available as a standalone enzyme, a complete reverse transcription kit or as a master mix with Oligo(dT) or Random Primers. RNasin(R) RNase Inhibitor protection included with Kit and Mixes
  • Sensitive transcription of both high-copy and low-copy messages
  • Transcribes short and long transcripts; processes through secondary structure

Specifications

Content And Storage −30 to −10°C
Description GoScript Reverse Transcriptase
Form Liquid
pH 7
Quantity 100 Reactions
For Use With (Application) cDNA Synthesis, RT-PCR
Includes GoScript reverse transcriptase, GoScript 5X reaction buffer, MgCl2

U.S. Patent 6,242,235, Australian Patent 761757, Canadian Patent 2,335,153 and other patents and patents pending.