missing translation for 'onlineSavingsMsg'
Learn More

Promega Apo-ONE™ Homogeneous Caspase-3/7 Assay Kit

Numéro de catalogue. PRG7792
Encompass
Modifier l'affichage
Click to view available options
Quantity:
1 mL
10 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRG7792 1 mL
PRG7790 10 mL
PRG7791 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PRG7792 Fournisseur Promega Code fournisseur G7792

Comprend

Apo-ONE Caspase Substrate Z-DEVD-R110, 100X; Apo-ONE Homogeneous Caspase Buffer

Highly sensitive, homogeneous fluorescent apoptosis assay for use in cultured cells and with purified caspase. Just add-mix-read.

  • Fast and Sensitive Measurement of Active Caspase-3 and -7
  • Fluorescent add-mix-read apoptosis assay for use in cultured cells and with purified caspase
  • Dramatically decrease time to results with highly sensitive and simple protocol
  • Flexible assay design allows for adaptable formats and throughput needs

Spécifications

Quantity 1 mL
Format Cuvettes, 96- or 384-well Plates
Content And Storage 1 x 1mL 100X Caspase substrate Z-DEVD-R110, 1 x 1mL Apo-ONE Homogeneous Caspase-3/7 Buffer. Store at -30°C to -10°C
Detection Method Fluorescence