Learn More
Invitrogen™ Power Blotter Station DFS Item
Get rapid and efficient semi-dry protein transfer with the Invitrogen™ Power Blotter.
Supplier:  Invitrogen™ PB0010
Numéro de catalogue. PB0010
Il en reste null
Découvrez plus d'offres spéciales
Comprend: Software with step-by-step instructions for creating, running, and saving custom programs
Description
Description
The Invitrogen™ Power Blotter Station is a power supply with touchscreen user interface for semi-dry transfer of proteins from gel to membrane when used with Power Blotter Cassette or Power Blotter Cassette XL. Power Blotter Station features pre-programmed electrophoretic protein transfer methods for different types, numbers, and sizes of gels and molecular weight ranges of proteins.
  • Easy-touch programming
  • Intuitive, color LCD touchscreen interface
  • Pre-programmed methods for rapid semi-dry transfer of proteins
  • Flexible—efficiently transfer proteins in about 5-12 minutes with 1 system
  • Integrated power supply—seamless operation between control unit and cassettes enables consistent high efficiency protein staining and transfer
  • Convenient—simultaneous transfer up to 2 mini-sized gels or 1 midi-sized gel using the Power Blotter Cassette, or transfer 4 mini-sized gels or 2 midi-sized gels using the Power Blotter Cassette XL
Spécifications
Spécifications
Western Blot
Nitrocellulose, PVDF
Power Blotter
1 unit
PB0002 or PB0003 (casettes)
Semi-Dry
Blotter Station
Room Temperature
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.