missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, ACS Reagent, Solstice

Numéro de catalogue. 6003137
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
4 x 2.5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6003137 1 kg
6003141 500 g
6003138 2.5 kg
6003142 6 x 500 g
6003140 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 6003137 Fournisseur Solstice Code fournisseur P06621KG
Il en reste null

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Identifiants chimiques

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Spécifications

Quantity 1 kg
Linear Formula KH2PO4
UN Number NONH for all modes of transport
Formula Weight 136.09g/mol
Percent Purity ≥99%
Grade ACS Reagent
Density 2.338g/mL at 25°C
Chemical Name or Material Potassium phosphate monobasic
RTECSNumber TC6615500