missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals

Numéro de catalogue. ICN19472780
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN19472780 100 g
ICN19472790 500 g
ICN19472791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. ICN19472780 Fournisseur MP Biomedicals Inc Code fournisseur 0219472780
Il en reste null

Identifiants chimiques

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Spécifications

Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99+%
Formula Weight 136.09
Chemical Name or Material Potassium Phosphate Monobasic, Anhydrous
Recommended Storage Room Temperature