missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium phosphate, 0.5M buffer soln., pH 7.4

Numéro de catalogue. p-7042126
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ61413AK 250 mL
AAJ61413AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAJ61413AK Fournisseur Thermo Scientific Chemicals Code fournisseur J61413AK
Il en reste null

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol

Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium phosphate is used as a buffer for the determination of pH. It is also used in pharmaceutical production, urinary acidifier, paper processing, nutrient solutions and yeast foods.

Solubility
Miscible with water. Immiscible with alcohol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Identifiants chimiques

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Spécifications

Concentration 0.5 M
pH 7.4
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Potassium phosphate
Recommended Storage Ambient temperatures

RUO – Research Use Only