missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 20% (w/v) Aqueous Solution, Spectrum™ Chemical

Numéro de catalogue. 18612769
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18612769 500 mL Amber Glass Bottle
18612770 1 L Amber Glass Bottle
18612771 4 L Amber Glass Bottle
18612772 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18612769 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur P433500MLGL
Il en reste null

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.