missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical

Supplier:  Ricca Chemical Company 62801

 View more versions of this product

Catalog No. 6280-1


Only null left

Description

Description

Specifications

Specifications

Potassium Iodate-Iodide
7732-18-5 , 7758-05-6 , 7758-05-6 , 1310-73-2
100.0,0.44,0.05,0.0016
214 g/mol
Laboratory
Slightly Alkaline
Liquid
Colorless
98.0,0.42,0.05,0.0016
dihydrogen oxide, dihydrogen monoxide
1 mL = 0.5 mg SO
Amber Glass Bottle
4 L
0.0125Normal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions