CAS RN 7758-05-6
Potassium Iodate Volumetric Solution, 0.0500 M (M/20), 0.300 N, Ricca Chemical
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710
Potassium iodate, 0.1N Standardized Solution
Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 MDL Number: MFCD00011406 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
Potassium iodate, 99.5%, for analysis
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214 MDL Number: MFCD00011406 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M IUPAC Name: potassium iodate SMILES: [K+].[O-][I](=O)=O
Potassium iodate, 98%, extra pure
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214 MDL Number: MFCD00011406 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M IUPAC Name: potassium iodate SMILES: [K+].[O-][I](=O)=O
Potassium Iodate, 0.00564 N, 200 ppm as Cl2, Ricca Chemical
CAS: 7758-05-6
Potassium Iodate, 0.0250 N (N/40), Ricca Chemical
Chemical Name or Material: Potassium Iodate CAS: 7758-05-6 CAS Min %: 0.09 CAS Max %: 0.09 CAS: 7732-18-5 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 144-55-8 CAS Min %: 0.05 CAS Max %: 0.05
Potassium Iodate, 0.160 N, Ricca Chemical
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
Potassium Iodate, 0.100 N (N/10), 0.01667 M (M/60), Ricca Chemical
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
Potassium Iodate, 0.0500 N (N/20), Ricca Chemical
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium iodate SMILES: [K+].[O-][I](=O)=O
Potassium Iodate, Certified, 0.1250N ±0.0005N (0.0208M), LabChem™
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
Potassium Iodate, Certified, 0.0250N ±0.0002N (0.0042M), LabChem™
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
Potassium Iodate, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™
CAS: 7758-05-6 Molecular Formula: IKO3 Molecular Weight (g/mol): 214.00 InChI Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M Synonym: potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat PubChem CID: 23665710 IUPAC Name: potassium;iodate SMILES: [O-]I(=O)=O.[K+]
- 1
- 2