missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate-Iodide, 0.0125 N (N/80), 1 mL = 0.5 mg SO32-, Ricca Chemical

$185.43 - $1,104.50

Specifications

Grade Laboratory
pH Slightly Alkaline
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6280-16
View Documents
Ricca Chemical Company
628016
500 mL Amber Glass Bottle N/A
Only null left
 
6280-32
View Documents
Ricca Chemical Company
628032
1 L Amber Glass Bottle N/A
Only null left
 
6280-1
View Documents
Ricca Chemical Company
62801
4 L Amber Glass Bottle N/A
Only null left
 
R62800004F
View Documents
Ricca Chemical Company
R62800004F
4 L Cubitainer N/A
Only null left
 
6280-5
View Documents
Ricca Chemical Company
62805
20 L Cubitainer N/A
Only null left
 
Description

Description

Specifications

Specifications

Laboratory
Slightly Alkaline
Videos
SDS
Documents

Documents