missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, Certified, 1.000N ±0.005N (1.0M), LabChem™

Supplier:  LabChem LC193505

 Afficher plus de versions de ce produit

Numéro de catalogue. LC193505


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Potassium Hydroxide
1310-58-3,7732-18-5
HKO
MFCD00003553
UN1814
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
20 L
[OH-].[K+]
56.11
CHEBI:32035
Certified
Cubitainer™
Traceable to NIST
Colorless
94.5,5.5
KOH
1.000N ±0.005N (1.0M)
1.05g/mL
Soluble in water
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
56.11
Passes Test
1.05g/mL
Liquid

Identifiants chimiques

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

FDS
Documents

Documents

Promotions

Promotions