missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Porcine TGF alpha ELISA Kit

Numéro de catalogue. ES21RBX5
Encompass
Modifier l'affichage
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ES21RBX5 5 x 96 Tests
ES21RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. ES21RBX5 Fournisseur Invitrogen™ Code fournisseur ES21RBX5
Obtenez jusqu’à 3,000 $ de produits Extra Life Sciences avec achat
Afficher toutes les promotions

ELISA

Porcine TGF alpha quantitates porcine TGF alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine TGF alpha.

TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Spécifications

Accession Number EGF-like TGF; ETGF; LOC100009482; protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; Tgfa; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1
Assay Range 0.6 to 120 pg/mL
Assay Sensitivity 0.6 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 397484
Gene Symbol TGFA
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TGF alpha
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Afficher plus de résultats Afficher moins de résultats