missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ QS, 80-100 mesh

Numéro de catalogue. 111004047
Modifier l'affichage
Click to view available options
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
111004047 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111004047 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur SGMA20343
Il en reste null

Porapak™™ QS packed columns maybe used for the analysis of sulphur-containing gases by gas-solid chromatograpy.

Porapak™ porous polymers are spherical beads with consistent particle size, porosity, and surface area that help to insure analytical precision and accuracy.

Spécifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜550 m2/g
Product Line PoraPak
For Use With (Application) GC
Density ˜0.34 g/mL (free fall density)

PoraPak is a trademark of Waters