missing translation for 'onlineSavingsMsg'
Learn More

POPSO, 0.2M buffer soln., pH 8.0, Thermo Scientific™

Numéro de catalogue. p-7041742
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ60085AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AAJ60085AK Fournisseur Thermo Scientific Chemicals Code fournisseur J60085AK
Il en reste null

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 108321-07-9
Molecular Formula C10H20N2Na2O8S2
Molecular Weight (g/mol) 406.376
MDL Number MFCD00070005
InChI Key XMRCLLXGMGEVHS-UHFFFAOYSA-L
PubChem CID 53393457
IUPAC Name disodium;2-hydroxy-3-[4-(2-hydroxy-3-sulfonatopropyl)piperazin-1-yl]propane-1-sulfonate
SMILES C1CN(CCN1CC(CS(=O)(=O)[O-])O)CC(CS(=O)(=O)[O-])O.[Na+].[Na+]

Spécifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material POPSO buffer soln.
TSCA No
Recommended Storage Keep cold