missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polystyrene Sign: CAUTION - SAFETY GLASSES REQUIRED BEYOND THIS POINT
Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$13.59 - $20.08
Spécifications
| Material | Polystyrene |
|---|---|
| Shape | Rectangular |
| Displayed Text | CAUTION - SAFETY GLASSES REQUIRED BEYOND THIS POINT |
| Color | Black on Yellow |
| Numéro de catalogue | Numéro du manufacturier. | Dimensions (L x W) | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Dimensions (L x W) | Prix | Quantité | |||||
|
19-111-771
|
Brady™
22593 |
14 x 10 in. |
chaque for $20.08
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
19-111-770
|
Brady™
22592 |
10 x 7 in. |
chaque for $13.59
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Polystyrene material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Polystyrene signs are thin, pliable and durable
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Spécifications
| Polystyrene | |
| Rectangular | |
| CAUTION - SAFETY GLASSES REQUIRED BEYOND THIS POINT | |
| Black on Yellow |