Learn More
PolyCo™ VR™ Protective Ankle-High Shoe Covers
Designed for comfort, flexibility and protection
Supplier:  PolyCo™ 49315
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-125-541
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Choice of 4mil or 6mil thickness
  • Water fluid resistance
  • Chemical resistance
  • Elastic top for added splash protection
  • Gritted material surface for slip resistance
  • Made in USA
Spécifications
Spécifications
Regular
Yellow
Heat sealed
Ideal for use where biological and chemical liquids are present
Polyolefin
Elastic Top
6 mil
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS