Learn More
Poltex Three Compartment Benchtop Organizer
Catalog No. 22000161
Encompass
Change view
Click to view available options
Total Positions:
3
Catalog No. Total Positions
22000161 3
1 options
Catalog No. 22000161 Supplier Poltex Supplier No. GBKIMTRIO
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Dispenser helps organize common laboratory items that typically do not have a permanent space on the benchtop. Designed to hold two boxes of disposable gloves, two boxes of lab wipes, with a slide cutter for a roll of parafilm.
  • Constructed from durable, break-resistant 1/4 in. PETG
  • Holds two boxes of disposable gloves, two boxes of lab wipes, and one roll of parafilm
  • Base includes rubber bump-ons to minimize slipping and scratching
  • Consumables are shown for demonstration purposes only
Specifications
Product Type Deluxe Benchtop Organizer
Includes Non-slick rubber bump-ons
Holds Disposable Glove Boxes, Labwipes, Parafilm
Total Positions 3
Dimensions (D x W x H) Interior 6 x 11 x 10.5 in.
Color Clear
Material PETG