Learn More
United Scientific™ Pipette Pump Set of 3
Convenient set of three pipette pumps.
Supplier:  United Scientific™ PPMPSET3
Numéro de catalogue. S03077
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
High quality pipetters designed for easy and precise pipetting with one hand. Pumps feature a lever for quick release, and comfortable forward or backward wheel movement. The soft elastic chuck has a threaded collar to tightly grip glass or plastic pipettes. Easy to clean and maintain.
  • The set consists of three sizes and colors: 1 Ea. Blue (2 mL), 1 Ea. Green (10 mL), 1 Ea. Red (25 mL)
  • Designed for repeated use.
  • Easy to care for and clean.
  • Compatible with plastic and glass pipettes
  • Smooth thumb wheel action for one hand operation
Spécifications
Spécifications
No
Assorted
Serologic Pipette
Plastic
1
Manual Pipette
Not Autoclavable
Reusable
Ergonomic
Junior
Non-sterile
Various
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS