missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol TS, 2% (w/v) Alcoholic, Ricca Chemical

$219.77 - $219.77

Chemical Identifiers

CAS 6099-90-7
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol dihydrate
SMILES O.O.OC1=CC(O)=CC(O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5775-16
View Documents
Ricca Chemical Company
577516
500 mL Amber Glass Bottle
Each for $219.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6099-90-7
162.14
phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
benzene-1,3,5-triol dihydrate
C6H10O5
MPYXTIHPALVENR-UHFFFAOYSA-N
80196
O.O.OC1=CC(O)=CC(O)=C1

Specifications

Brown-Yellow to Colorless
2.39,86.0,4.57
C6H10O5
MPYXTIHPALVENR-UHFFFAOYSA-N
benzene-1,3,5-triol dihydrate
80196
Laboratory
Alcoholic Solution
Liquid
Phloroglucinol, Test Solution
6099-90-7 , 67-63-0 , 64-17-5
2.49,90.0,4.75
phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
O.O.OC1=CC(O)=CC(O)=C1
162.14
2%
Amber Glass Bottle
500 mL
2% (w/v)
Videos
SDS
Documents

Documents