missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucinol TS, 2% (w/v) Alcoholic, Ricca Chemical

Catalog No. 577516
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
577516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 577516 Supplier Ricca Chemical Company Supplier No. 577516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6099-90-7 , 67-63-0 , 64-17-5
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
InChI Key MPYXTIHPALVENR-UHFFFAOYSA-N
Synonym phloroglucinol dihydrate, benzene-1,3,5-triol dihydrate, 1,3,5-trihydroxybenzene dihydrate, 1,3,5-benzenetriol, dihydrate, phloroglucinol, dihydrate, sym-trihydroxybenze, unii-0x7b22brg1, ccris 6521, 1,3,5-benzoltrioldihydrat, phloroglucinoldihydrate
PubChem CID 80196
IUPAC Name benzene-1,3,5-triol dihydrate
SMILES O.O.OC1=CC(O)=CC(O)=C1

Specifications

Color Brown-Yellow to Colorless
CAS Min % 2.39, 86.0, 4.57
CAS Max % 2.49, 90.0, 4.75
Percent Purity 2%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Alcoholic Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2% (w/v)
Chemical Name or Material Phloroglucinol, Test Solution
Show More Show Less