accessibility menu, dialog, popup

UserName

H2OH2OOHOHHO

CAS RN 6099-90-7

IUPAC Name: benzene-1,3,5-triol dihydrate
Synonyms: phloroglucinol dihydrate
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: MPYXTIHPALVENR-UHFFFAOYSA-N
SMILES: O.O.OC1=CC(O)=CC(O)=C1
Molecular Weight (g/mol) 
  • (4)
  • (7)
Quantity 
  • (4)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (5)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (4)
  • (7)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)