Learn More
Phenylarsine Oxide, (1mL = 0.2mg Cl), Certified, 0.00564N ±0.00005N (0.00282M), LabChem™
Supplier: LabChem LC183602
Spécifications
| Phenylarsine Oxide | |
| 637-03-6,7732-18-5 | |
| Colorless | |
| Amber Glass | |
| C6H5AsO | |
| 1 L | |
| Soluble in water | |
| O=[As]C1=CC=CC=C1 | |
| 168.03 | |
| CHEBI:75253 | |
| Certified | |
| Liquid |
| (1mL = 0.2mg Cl) | |
| 99.9,0.1 | |
| 6.0 to 7.0 | |
| C6H5AsO | |
| MFCD00001990 | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| arsorosobenzene | |
| 4778 | |
| 168.03 | |
| 0.00564N ±0.00005N (0.00282M) |
Identifiants chimiques
| 637-03-6 | |
| 168.03 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| 4778 | |
| arsorosobenzene |
| C6H5AsO | |
| MFCD00001990 | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| CHEBI:75253 | |
| O=[As]C1=CC=CC=C1 |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature