Learn More
Micro Essential Lab pH Test Paper Refills for Hydrion™ Dispensers
Hydrion™ pH Paper provides excellent color separation in the alkaline range and offers clear bright single color matches for various pH ranges.
$76.13 - $102.61
Specifications
| For Use With (Equipment) | For Hydrion™ dispensers |
|---|---|
| Includes | pH paper, Matching color chart |
| No. of Tests | 5000 Tests |
| Quantity | 5 Pk. |
| Dimensions (L x W) | 4.6 m x 0.55 cm (15 ft. x 0.21 in.) |
| Catalog Number | Mfr. No. | pH Range | Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | pH Range | Type | Price | Quantity & Availability | ||||
|
1485350
|
Micro Essential Lab
MES405 |
0 to 3 | Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
1485360
|
Micro Essential Lab
MES415 |
1 to 2.5 | Short-range pH Test Paper |
Pack of 5 Each for $90.48
Case of 10 Pack for $904.80
|
|
||||
|
1485320
|
Micro Essential Lab
1601 6 |
1 to 6 | Wide-range pH Test Paper |
Pack of 5 Each for $76.13
Case of 10 Pack for $761.30
|
|
||||
|
14853B
|
Micro Essential Lab
160B |
1 to 11 | Wide-range pH Test Paper |
Pack of 5 Each for $84.29
Case of 10 Pack for $842.90
|
|
||||
|
1485327
|
Micro Essential Lab
160/111VIV |
1 to 11 | Vivid/Wide-range pH Test Paper |
Pack of 5 Each for $90.48
Case of 10 Pack for $904.80
|
|
||||
|
1485322
|
Micro Essential Lab
1601 12 |
1 to 12 | Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
14853A
|
Micro Essential Lab
160A |
2 to 10 | Wide-range pH Test Paper |
Pack of 5 Each for $102.61
Case of 10 Pack for $1,026.10
|
|
||||
|
1485370
|
Micro Essential Lab
MES425 |
3 to 5.5 | Short-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
1485323
|
Micro Essential Lab
1603 9 |
3 to 9 | Jumbo/Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
1485324
|
Micro Essential Lab
160/49 |
4 to 9 | Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
1485390
|
Micro Essential Lab
MES445 |
6 to 8 | Wide-range pH Test Paper |
Pack of 5 Each for $83.10
Case of 10 Pack for $831.00
|
|
||||
|
1485325
|
Micro Essential Lab
1606 11 |
6 to 11 | Wide-range pH Test Paper |
Pack of 5 Each for $89.74
Case of 10 Pack for $897.40
|
|
||||
|
14853103
|
Micro Essential Lab
458 |
8 to 9.5 | Short-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
14853112
|
Micro Essential Lab
467 |
9 to 12 | Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
14853120
|
Micro Essential Lab
475 |
10 to 12 | Wide-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
|
14853140
|
Micro Essential Lab
495 |
12.5 to 14 | Short-range pH Test Paper |
Pack of 5 Each for $80.84
Case of 10 Pack for $808.40
|
|
||||
Description
Hydrion™ pH Paper is a general use pH paper. A sample of its diverse pH applications include:
- Classroom Demonstrations
- Hospital Laboratories
- General Laboratories
- Industrial Testing
- Food Service
- Safety Testing
- Emergency Testing
- Agricultural Testing
Each kit contains 5, 15-foot roll of test paper and matching color chart. Provides for approximately 5000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Specifications
| For Hydrion™ dispensers | |
| 5000 Tests | |
| 4.6 m x 0.55 cm (15 ft. x 0.21 in.) | |
| 4.6 m | |
| 0.55 cm |
| pH paper, Matching color chart | |
| 5 Pk. | |
| 15 ft. | |
| 0.21 in. |
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.