Learn More
Micro Essential Lab pH Test Paper Refills for Hydrion™ Dispensers
Hydrion™ pH Paper provides excellent color separation in the alkaline range and offers clear bright single color matches for various pH ranges.
Supplier: Micro Essential Lab MES415
Description
Hydrion™ pH Paper is a general use pH paper. A sample of its diverse pH applications include:
- Classroom Demonstrations
- Hospital Laboratories
- General Laboratories
- Industrial Testing
- Food Service
- Safety Testing
- Emergency Testing
- Agricultural Testing
Each kit contains 5, 15-foot roll of test paper and matching color chart. Provides for approximately 5000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Chemical Identifiers
| For Hydrion™ dispensers | |
| 5000 Tests | |
| 4.6 m x 0.55 cm (15 ft. x 0.21 in.) | |
| 4.6 m | |
| 0.55 cm |
| pH paper, Matching color chart | |
| 5 Pk. | |
| 15 ft. | |
| 0.21 in. |
Specifications
| 1 to 2.5 | |
| pH paper, Matching color chart | |
| 5 Pk. | |
| 4.6 m x 0.55 cm (15 ft. x 0.21 in.) | |
| 4.6 m | |
| 0.55 cm |
| For Hydrion™ dispensers | |
| 5000 Tests | |
| Short-range pH Test Paper | |
| 15 ft. | |
| 0.21 in. |
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.