Learn More
Micro Essential Lab pH Test Paper Refills for Hydrion™ Dispensers
Hydrion™ pH Paper provides excellent color separation in the alkaline range and offers clear bright single color matches for various pH ranges.
Supplier:  Micro Essential Lab MES425
 View more versions of this product
Catalog No. 1485370
Only null left
Description
Description
Hydrion™ pH Paper is a general use pH paper. A sample of its diverse pH applications include:
  • Classroom Demonstrations
  • Hospital Laboratories
  • General Laboratories
  • Industrial Testing
  • Food Service
  • Safety Testing
  • Emergency Testing
  • Agricultural Testing
Each kit contains 5, 15-foot roll of test paper and matching color chart. Provides for approximately 5000 tests.
Directions
Simply tear off a small strip of pH paper, dip into the test solution, then instantly compare the resulting color with the matching pH color chart.
Specifications
Chemical Identifiers
For Hydrion™ dispensers
5000 Tests
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
pH paper, Matching color chart
5 Pk.
15 ft.
0.21 in.
Specifications
3 to 5.5
pH paper, Matching color chart
5 Pk.
4.6 m x 0.55 cm (15 ft. x 0.21 in.)
4.6 m
0.55 cm
For Hydrion™ dispensers
5000 Tests
Short-range pH Test Paper
15 ft.
0.21 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
This product is not intended to diagnose, treat, cure or prevent any disease or condition. Please contact a medical doctor to diagnose and treat any medical condition.