Learn More
VeriSpec™ pH Buffer, Certified Reference Material
ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier: Ricca VeriSpec™ RV159000500A
Description
- No formaldehyde or mercury compounds as preservatives
- No chromium compounds as colorants
- Manufactured and tested in ISO 17025/17034 accredited facilities
- pH 4, 7, and 10 are color coded for easy identification
- Certified traceable to NIST
Specifications
| 7732-18-5 , 7447-40-7 , 1310-73-2 , 10043-35-3 | |
| Yes | |
| pH Meter and Electrode | |
| Liquid | |
| 500 mL | |
| 500 mL Natural Poly | |
| Certified |
| Colorless | |
| Calibration | |
| 9.00 ±0.01 | |
| VeriSpec | |
| pH Buffer | |
| ISO 17025/17034 |
ISO 17025/ISO 17034