missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VeriSpec™ pH Buffer, Certified Reference Material
ISO 17025/17034 pH Buffer Certified Reference Materials provide the highest level of quality assurance. A certificate of analysis providing the specified property value, uncertainty, and traceability is available with every bottle.
Supplier: VeriSpec™ RV1601001A
Description
- No formaldehyde or mercury compounds as preservatives
- No chromium compounds as colorants
- Manufactured and tested in ISO 17025/17034 accredited facilities
- pH 4, 7, and 10 are color coded for easy identification
- Certified traceable to NIST
Specifications
| VeriSpec™ Certified Reference Material, Buffer pH 10.00 ±0.01 (at 25°C) | |
| Blue | |
| 1X | |
| pH Meter and Electrode | |
| Liquid | |
| 1 L | |
| Aqueous Solution | |
| NIST Traceable |
| Proprietary , 497-19-8 , 1310-73-2 , 144-55-8 , 7732-18-5 | |
| Yes | |
| Calibration | |
| 10.00 ±0.01 | |
| VeriSpec | |
| pH Buffer | |
| Natural Poly | |
| Certified |
ISO 17025/ISO 17034