missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Honeywell Riedel-de Haën™
CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons),≥99.0%
$485.20 - $485.20
Identifiants chimiques
| CAS | 109-66-0 |
|---|---|
| Molecular Formula | C5H12 |
| Molecular Weight (g/mol) | 72.15 |
| MDL Number | MFCD00009498 |
| InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
| Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
| PubChem CID | 8003 |
| ChEBI | CHEBI:37830 |
| IUPAC Name | pentane |
| SMILES | CCCCC |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
6001121
|
Honeywell-Chromasolv
768661L |
1 L |
chaque for $485.20
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Identifiants chimiques
| 109-66-0 | |
| 72.15 | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| 8003 | |
| pentane |
| C5H12 | |
| MFCD00009498 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| CHEBI:37830 | |
| CCCCC |
Spécifications
| 109-66-0 | |
| 1 L | |
| −49°C (−56.2°F) | |
| C5H12 | |
| MFCD00009498 | |
| 969132 | |
| OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
| pentane | |
| 8003 | |
| 72.15g/mol | |
| CHROMASOLV |
| 0.626g/mL at 25°C | |
| 0.083 | |
| 500°F | |
| CH3(CH2)3CH3 | |
| UN1265 | |
| n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
| CCCCC | |
| 72.15 | |
| CHEBI:37830 | |
| ≥99%, ≥99% (GC) | |
| Pentane |
Sécurité et manipulation
P210-P261-P273-P301 + P310-P331
missing translation for 'rtecsNumber' : RZ9450000