Learn More
Cytiva (Formerly GE Healthcare Life Sciences) PBS-P + Buffer 10 x
Numéro de catalogue. 501055502
Change view
Click to view available options
PBS-P + Buffer 10 x
Numéro de catalogue.
501055502 PBS-P + Buffer 10 x
1 options
Numéro de catalogue. 501055502 Fournisseur Cytiva (Formerly GE Healthcare Life Sciences) Code fournisseur 28995084
Il en reste null
Phosphate buffered saline containing detergent used as running buffer for small molecule analysis
  • Features:
    • Concentrated stock solution, yielding pH 7.4 when diluted 10 x and supplemented with 2% DMSO
    Uses:
    • Used in Biacore systems
    • PBS buffer with DMSO commonly used for small molecule compound samples
Spécifications
For Use With (Application) Analytical Research, Research, Interaction Analysis in Biacore Systems, Small Molecule Assays
For Use With (Equipment) All Biacore SPR Systems
Quantity 1000 mL