missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth LDPE Reagent Bottles with Polypropylene Caps
Pack of 12 wide-mouth LDPE reagent bottles with polypropylene caps are non-sterile, leak-tested, and chemically resistant for safe, easy handling and storage of lab liquids in research and industrial settings.
Supplier: Eisco Labs CH5516PK12
Description
- Durable LDPE construction: Pack of 12 reagent bottles made from LDPE with polypropylene caps. Designed for reliable chemical resistance and safe liquid storage.
- Wide mouth design: Wide-mouth opening ensures easy filling, pouring, and handling, ideal for laboratory and industrial applications.
- Chemical resistance: Resistant to a variety of chemicals, making it suitable for diverse uses in laboratories, research, and industrial environments. Non-sterile and non-autoclavable, with a temperature range of -100°C to +80°C.
- Leakproof testing: Bottles are tested for leakage under high pressure, ensuring safe storage and transportation of liquids and reagents.
- Translucent and lightweight: Translucent material provides visibility of contents, while lightweight construction ensures ease of carrying and handling.
Chemical Identifiers
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| LDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Specifications
| 60 mL | |
| Translucent | |
| LDPE | |
| Non-sterile | |
| No | |
| 85 mm | |
| 39.8 mm |
| 2 oz. | |
| 21.6 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Not Autoclavable |