missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth LDPE Reagent Bottles with Polypropylene Caps
Pack of 12 wide-mouth LDPE reagent bottles with polypropylene caps are non-sterile, leak-tested, and chemically resistant for safe, easy handling and storage of lab liquids in research and industrial settings.
Specifications
| Color | Translucent |
|---|---|
| Material | LDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000133
|
Eisco Labs
CH5515PK12 |
30 mL |
N/A
|
|
|||||
|
S000134
|
Eisco Labs
CH5516PK12 |
60 mL |
N/A
|
|
|||||
|
S000135
|
Eisco Labs
CH5518PK12 |
250 mL |
N/A
|
|
|||||
|
S000136
|
Eisco Labs
CH5519PK12 |
500 mL |
N/A
|
|
|||||
|
S000137
|
Eisco Labs
CH5520PK12 |
1 L |
N/A
|
|
|||||
Description
- Durable LDPE construction: Pack of 12 reagent bottles made from LDPE with polypropylene caps. Designed for reliable chemical resistance and safe liquid storage.
- Wide mouth design: Wide-mouth opening ensures easy filling, pouring, and handling, ideal for laboratory and industrial applications.
- Chemical resistance: Resistant to a variety of chemicals, making it suitable for diverse uses in laboratories, research, and industrial environments. Non-sterile and non-autoclavable, with a temperature range of -100°C to +80°C.
- Leakproof testing: Bottles are tested for leakage under high pressure, ensuring safe storage and transportation of liquids and reagents.
- Translucent and lightweight: Translucent material provides visibility of contents, while lightweight construction ensures ease of carrying and handling.
Specifications
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| LDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |