missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
Store and handle lab liquids with confidence using Eisco Labs wide-mouth translucent HDPE bottles, each fitted with a polypropylene cap for leakproof performance, chemical resistance, and easy pouring in demanding lab or industrial settings.
Supplier: Eisco Labs CH5465PK12
Description
- Reagent bottles: Includes twelve translucent wide-mouth bottles made of HDPE, featuring polypropylene caps, ideal for reagent storage and lab use.
- Excellent chemical resistance: Made with HDPE material that resists a wide variety of chemicals, ensuring reliable containment for diverse applications in laboratories and industries.
- Temperature resistance: Functional in a range from -100°C to 120°C, these bottles are designed for safe handling of solutions in cold and moderately heated conditions.
- Leakproof design: Rigorously tested to prevent leaks, these bottles offer secure transportation and storage for liquids and reagents.
- Ergonomic and translucent: Wide-mouth design ensures easy filling and pouring. Translucent material allows visibility of contents for efficient handling.
Identifiants chimiques
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Spécifications
| 500 mL | |
| Translucent | |
| HDPE | |
| Non-sterile | |
| No | |
| 167 mm | |
| 73.7 mm |
| 17 oz. | |
| 43.7 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Not Autoclavable |