missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth HDPE Reagent Bottles with Polypropylene Caps
Store and handle lab liquids with confidence using Eisco Labs wide-mouth translucent HDPE bottles, each fitted with a polypropylene cap for leakproof performance, chemical resistance, and easy pouring in demanding lab or industrial settings.
$52.39 - $209.43
Specifications
| Color | Translucent |
|---|---|
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000117
|
Eisco Labs
CH5462PK12 |
60 mL |
Pack of 12 Each for $59.36
|
|
|||||
|
S000118
|
Eisco Labs
CH5463PK12 |
125 mL |
Pack of 12 Each for $69.82
|
|
|||||
|
S000119
|
Eisco Labs
CH5464PK12 |
250 mL |
Pack of 12 Each for $97.71
|
|
|||||
|
S000120
|
Eisco Labs
CH5465PK12 |
500 mL |
Pack of 12 Each for $139.61
|
|
|||||
|
S000121
|
Eisco Labs
CH5466PK12 |
1 L |
Pack of 12 Each for $209.43
|
|
|||||
|
S000116
|
Eisco Labs
CH5461PK12 |
30 mL |
Pack of 12 Each for $52.39
|
|
|||||
Description
- Reagent bottles: Includes twelve translucent wide-mouth bottles made of HDPE, featuring polypropylene caps, ideal for reagent storage and lab use.
- Excellent chemical resistance: Made with HDPE material that resists a wide variety of chemicals, ensuring reliable containment for diverse applications in laboratories and industries.
- Temperature resistance: Functional in a range from -100°C to 120°C, these bottles are designed for safe handling of solutions in cold and moderately heated conditions.
- Leakproof design: Rigorously tested to prevent leaks, these bottles offer secure transportation and storage for liquids and reagents.
- Ergonomic and translucent: Wide-mouth design ensures easy filling and pouring. Translucent material allows visibility of contents for efficient handling.
Specifications
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |