Learn More
Promega P450-Glo™ CYP3A4 Assay with Luciferin-IPA
The P450-Glo CYP3A4 Assay System provides a homogeneous, luminescent method for measuring CYP3A4 activity and the screening system contains a complete set of reagents for performing the assay.
Supplier:  Promega V9002
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRV9002
Il en reste null
Comprend: Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
Description
Description
  • Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
  • Screening systems include complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background
Spécifications
Identifiants chimiques
-30°C to -10°C
Homogeneous, Luminescence
For measuring cytochrome P450 activity
P450-Glo CYP3A4 Assay with Luciferin-IPA
Spécifications
50 mL
Homogeneous, Luminescence
1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer with Esterase, 1 x 60μL Luciferin-IPA
-30°C to -10°C
For measuring cytochrome P450 activity
P450-Glo CYP3A4 Assay with Luciferin-IPA
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS