missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORP Standard, 200 mV vs. Ag/AgCl, Reagents
ORP Standard, 200 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CO0002001C
Specifications
| ORP Standard, 200 mV vs. Ag/AgCl | |
| 7732-18-5 , 7447-40-7 , 14459-95-1 | |
| Liquid | |
| Yellow/Green | |
| Oxidation Reduction Potential Standard | |
| Completely miscible | |
| Reagent |
| Glass Bottle | |
| 1.0052 | |
| Redox | |
| 100°C | |
| Redox Meter | |
| 1 L |