missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, 200 mV vs. Ag/AgCl, Reagents

Catalog No. p-200022039
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334589 10 L
01334588 1 L
01334587 20 L
01334586 4 L
01334585 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01334589 Supplier Labchem Inc Supplier No. CO00020010F
Only null left

ORP Standard, 200 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material ORP Standard, 200 mV vs. Ag/AgCl
Packaging Cubitainer
CAS 7732-18-5, 7447-40-7, 14459-95-1
Specific Gravity 1.0052
Physical Form Liquid
Technique Redox
Color Yellow/Green
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 10 L
Grade Reagent
Show More Show Less