missing translation for 'onlineSavingsMsg'
Learn More

ORP Standard, 200 mV vs. Ag/AgCl, Reagents

ORP Standard, 200 mV vs. Ag/AgCl, Reagent, use for Redox. Store in a Cool Dry Place. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CO0002004C

 View more versions of this product

Catalog No. 01-334-586


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

ORP Standard, 200 mV vs. Ag/AgCl
7732-18-5 , 7447-40-7 , 14459-95-1
Liquid
Yellow/Green
Oxidation Reduction Potential Standard
Completely miscible
Reagent
Glass Bottle
1.0052
Redox
100°C
Redox Meter
4 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions