Learn More
MilliporeSigma™ Supelco™ Omnifit™ EZ Benchmark Replacement Glass Columns
Replacement glass columns are available in a wide range of bore sizes and lengths.
Supplier:  MilliporeSigma™ Supelco™ K5170U
 View more versions of this product
Catalog No. 111009366
Only null left
Explore available promotions
Description
Description
Replacement columns are available to replace broken columns. The modular nature of Benchmark columns provides ultimate flexibility. Different column sizes can be mixed and matched with different endpiece types to meet specific requirements.
Specifications
Specifications
35 mm
150 mm
Omnifit
Chromatography
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Omnifit is a registered trademark of Diba Industries